Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Proteome Profiler Human XL Cytokine Array Kit
GSA_VAGREENER_CHOICE

Catalog No. ary022b
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ARY022B 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ARY022B Supplier R&D Systems™ Supplier No. ARY022B
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Proteome Profiler Human XL Cytokine Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.

The Proteome Profiler Human XL Cytokine Array Kit contains 4 membranes - each spotted in duplicate with 105 different cytokine antibodies.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Description Cytokine Assay Kits
Quantity 1 Kit
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Use With (Application) For parallel determination of human cytokines and chemokine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.