Learn More
R&D Systems™ Proteome Profiler Mouse Adipokine Array Kit
GREENER_CHOICESDP
Catalog No. ARY013 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
ARY013 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. ARY013 Supplier R&D Systems™ Supplier No. ARY013
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
The Proteome Profiler Mouse Adipokine Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Mouse Adipokine Array Kit contains 4 membranes - each spotted in duplicate with 38 different obesity-related antibodies.
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 Kit