Learn More
R&D Systems™ Proteome Profiler Mouse Chemokine Array Kit
GREENER_CHOICESDP
Catalog No. ARY020 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
ARY020 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. ARY020 Supplier R&D Systems™ Supplier No. ARY020
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
The Proteome Profiler Mouse Chemokine Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Mouse Chemokine Array Kit contains 4 membranes - each spotted in duplicate with 25 different chemokine antibodies.
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 Kit