Learn More
R&D Systems™ Proteome Profiler Mouse Phospho-RTK Array Kit
GREENER_CHOICESDP
Catalog No. ARY014 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
ARY014 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. ARY014 Supplier R&D Systems™ Supplier No. ARY014
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
The Proteome Profiler Mouse Phospho-RTK Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Mouse Phospho-RTK Array Kit contains 4 membranes - each spotted in duplicate with 39 different RTK antibodies.
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 Kit