Learn More
R&D Systems™ Proteome Profiler Rat Cytokine Array Kit, Panel A
GSA_VAGREENER_CHOICESDP
Catalog No. ARY008 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
ARY008 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. ARY008 Supplier R&D Systems™ Supplier No. ARY008
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
The Proteome Profiler Rat Cytokine Array Kit, Panel A requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Rat Cytokine Array Kit, Panel A contains 4 membranes - each spotted in duplicate with 29 different cytokine antibodies.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 Kit