Learn More
R&D Systems™ Proteome Purify 2 Human Serum Protein Immunodepletion Resin

Description
Includes
Immunodepletion Resin, Spin-X Filter Units
Allows user to increase protein load or concentration
Removal allows visualization of proteins that co-migrate or co-purify with the high-abundance proteins
Kit removes proteins representing approximately 60% of the protein contained in serum and plasma
Specifications
Specifications
| Quantity | 40 Tests |
| Product Type | Immunodepletion Resin |
| Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
| Product Line | R&D Systems |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.