Learn More
United States Biological Corporation PROTEOSE PEPTONENO3(PWDER)500G
Catalog No. NC9210308 Shop All United States Biological Corporation Products
Encompass_Preferred
Proteose Peptone, 3 Agar; United States Biological Corporation; Light beige, homogeneous, free-flowing powder, Solubility: Clear to sl. hazy, amber, complete w/heating; Moisture: 5% Ash: 12% Amino Nitrogen: 6% Total Nitrogen: 15% pH
Catalog No. NC9210308 Supplier United States Biological Corporation Supplier No. P911322500G
Proteose Peptone, 3 Agar; United States Biological Corporation; Light beige, homogeneous, free-flowing powder, Solubility: Clear to sl. hazy, amber, complete w/heating; Moisture: 5% Ash: 12% Amino Nitrogen: 6% Total Nitrogen: 15% pH