Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Proteus Monoclonal Antibody (31-17)
GREENER_CHOICE

Catalog No. 01672077 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-077 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-077 Supplier Invitrogen™ Supplier No. MA135772
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody reacts with bacteria in suspension and is therefore useful for detection of Proteus spp. in bacterial suspensions. Various systems can be used with this antibody to detect the presence of Proteus spp.. The antibody reacts with approximately 50% of the Proteus spp. isolates tested.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Proteus
Applications ELISA
Classification Monoclonal
Clone 31-17
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Host Species Mouse
Immunogen Proprietary immunogen.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.