Learn More
Micro Direct Inc PROTEX DISINFECTANT WIPE
SDP
Supplier:  Micro Direct Inc 4870
Encompass
The PROTEX disinfectant wipe is a one-step cleaner/disinfectant effective against a broad spectrum of pathogens including MRSA, HIV, Staph and many others. The 7"x10" textured wipe is supplied in a 75 count canister.
Catalog No. NC1380437