Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protocadherin-8 Antibody [PE], R&D Systems™
SDP

Catalog No. AF5434P
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5434P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AF5434P Supplier R&D Systems Supplier No. AF5434P

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

Protocadherin-8 Polyclonal specifically detects Protocadherin-8 in Human samples. It is validated for Immunocytochemistry, Western Blot.

Specifications

Antigen Protocadherin-8
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias ARCADLIN, PAPC, protocadherin 8, protocadherin-8
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Protocadherin-8, Lys30-Pro749, Accession # O95206
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5100
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.