Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PROTOCOL™ Histosolv-X, Fisher Healthcare

Catalog No. 23245686 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-245686 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-245686

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • D-limonene-based clearing agent that is safer to use than xylene
  • May be used as a safe and effective alternative to xylene in histology and cytology procedures

Chemical Identifiers

CAS 5989-27-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00062991
InChI Key XMGQYMWWDOXHJM-JTQLQIEISA-N
Synonym d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene
PubChem CID 440917
ChEBI CHEBI:15382
IUPAC Name (4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
SMILES CC(=C)[C@@H]1CCC(C)=CC1

Specifications

Quantity 4 L
Chemical Name or Material PROTOCOL™ Histosolv-X
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.