Learn More
Fisher Healthcare™ PROTOCOL™ Parasitology Single-Vial Kits, Clean Vials
Shop All Fisherbrand ProductsDescription
Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens with Thermo Scientific™ PROTOCOL™ Parasitology Single-Vial Kits.
Zinc Sulfite-Polyvinyl Alcohol (Zn-PVA)
- Mercury-free fixative for use as a preservative/fixative for protozoan trophozoites
- Slides fixed with Zn-PVA may be stained using trichrome or hematoxylin
Sodium Acetate Acetic Acid Formalin (SAF)
- Mercury-free preservative/fixative ideally suited for use with helminth eggs and larvae, protozoan trophozoites and cysts, coccidian oocysts and microsporidian spores
- May be stained using trichrome or hematoxylin
10% Neutral Buffered Formalin
- All-purpose fixative appropriate for helminth eggs and larvae, and for protozoan cysts
- Buffering system helps to maintain organism morphology
- Fixed specimens may be examined directly or concentrated for recovery of eggs, larvae and protozoan cysts
Modified Cupric Sulfate-Polyvinyl Alcohol (Modified Cu-PVA)
- Acts as a preservative/fixative for protozoan trophozoites
- Slides fixed with Modified PVA may be stained using trichrome or hematoxylin
Culture and Sensitivity (C & S Media)
- For collecting and transporting enteric bacterial pathogens such as Yersinia, Salmonella, Shigella, Campylobacter and Cholera Vibrio
- Bacterial population is maintained for optimum recovery, up to 96 hours
Cary-Blair Media
- Maintains the bacterial population of enteric pathogens such as Yersinia, Salmonella, Shigella, Campylobacter and Cholera Vibrio for up to 96 hours
Clean Vials
- Empty, 30mL vials for transporting specimens
Ordering Alerts:
Supplied as pack of 20 vials
Note:
All PROTOCOL Parasitology products are mercury free.
Specifications
Specifications
| Product Type | Parasitology Single Vial Kit |
| Media Type | Modified (Cu) PVA |
| Quantity | 20 Vials/Pk. |
| Volume (English) | 1.01 oz. |
| Volume (Metric) | 30 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.