Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ PROTOCOL™ Parasitology Single-Vial Kits, Clean Vials

Catalog No. 2300537 Shop All Fisherbrand Products
Change view
Click to view available options
Media Type:
10% Buffered Formalin
C&S
Cary-Blair
Clean
Modified (Cu) PVA
SAF
Zn-PVA
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type
23-005-37 Zn-PVA
23-005-46 10% Buffered Formalin
23-005-43 C&S
23-005-33 Modified (Cu) PVA
23-005-41 SAF
23-005-31 Clean
23-005-47 Cary-Blair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 23-005-37
Add to Cart
Add to Cart
This item is not returnable. View return policy

Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens.

Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens with Thermo Scientific™ PROTOCOL™ Parasitology Single-Vial Kits.

Zinc Sulfite-Polyvinyl Alcohol (Zn-PVA)

  • Mercury-free fixative for use as a preservative/fixative for protozoan trophozoites
  • Slides fixed with Zn-PVA may be stained using trichrome or hematoxylin

Sodium Acetate Acetic Acid Formalin (SAF)

  • Mercury-free preservative/fixative ideally suited for use with helminth eggs and larvae, protozoan trophozoites and cysts, coccidian oocysts and microsporidian spores
  • May be stained using trichrome or hematoxylin

10% Neutral Buffered Formalin

  • All-purpose fixative appropriate for helminth eggs and larvae, and for protozoan cysts
  • Buffering system helps to maintain organism morphology
  • Fixed specimens may be examined directly or concentrated for recovery of eggs, larvae and protozoan cysts

Modified Cupric Sulfate-Polyvinyl Alcohol (Modified Cu-PVA)

  • Acts as a preservative/fixative for protozoan trophozoites
  • Slides fixed with Modified PVA may be stained using trichrome or hematoxylin

Culture and Sensitivity (C & S Media)

  • For collecting and transporting enteric bacterial pathogens such as Yersinia, Salmonella, Shigella, Campylobacter and Cholera Vibrio
  • Bacterial population is maintained for optimum recovery, up to 96 hours

Cary-Blair Media

  • Maintains the bacterial population of enteric pathogens such as Yersinia, Salmonella, Shigella, Campylobacter and Cholera Vibrio for up to 96 hours

Clean Vials

  • Empty, 30mL vials for transporting specimens

Ordering Alerts:

Supplied as pack of 20 vials

Note:

All PROTOCOL Parasitology products are mercury free.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Parasitology Single Vial Kit
Media Type Zn-PVA
Quantity 20 Vials/Pk.
Volume (English) 1.01 oz.
Volume (Metric) 30 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.