Learn More
Fisher Healthcare™ PROTOCOL™ Parasitology Two-Vial Kits, Modified Cu-PVA/10% Formalin Kits
Catalog No. 2300539 Shop All Fisherbrand Products
Change view
Click to view available options
Media Type:
Modified (Cu) PVA, Formalin
SAF, C&S
SAF, Clean
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type
23-005-39 SAF, Clean
23-005-27 SAF, C&S
23-005-45 Modified (Cu) PVA, Formalin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-005-39
Add to Cart
Add to Cart
Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens.
Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens with Thermo Scientific™ PROTOCOL™ Parasitology Two-Vial Kits.
Zinc Sulfite-Polyvinyl Alcohol (Zn-PVA)
  • Mercury-free fixative for use as a preservative/fixative for protozoan trophozoites
  • Slides fixed with Zn-PVA may be stained using trichrome or hematoxylin
Sodium Acetate Acetic Acid Formalin (SAF)
  • Mercury-free preservative/fixative ideally suited for use with helminth eggs and larvae, protozoan trophozoites and cysts, coccidian oocysts and microsporidian spores
  • May be stained using trichrome or hematoxylin
10% Buffered Formalin
  • All-purpose fixative appropriate for helminth eggs and larvae and for protozoan cysts
  • Buffering system helps to maintain organism morphology
  • Fixed specimens may be examined directly or concentrated for recovery of eggs, larvae and protozoan cysts
Modified Cupric Sulfate-Polyvinyl Alcohol (Modified Cu-PVA)
  • Acts as a preservative/fixative for protozoan trophozoites
  • Slides fixed with Modified PVA may be stained using trichrome or hematoxylin
Culture & Sensitivity (C & S Media)
  • For collecting and transporting enteric bacterial pathogens such as Yersinia, Salmonella, Shigella, Campylobacter, and CholeraVibrio
  • Bacterial population is maintained for optimum recovery, up to 96 hours
Clean Vials
  • Empty, 30mL vials for transporting specimens
Ordering Alerts:
Supplied as 10 packs of two vials each
Note:
All PROTOCOL Parasitology products are mercury free.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Parasitology Two Vial Kit
Description SAF/C and S Kits
Media Type SAF, Clean
Quantity 2 Pk, 10 Pk./Cs.
Volume (English) 1.01 oz.
Volume (Metric) 30 mL