Learn More
MBL International Corporation Anti-Proton Pump (H, K-ATPasebeta subunit) (Mouse) mAb (Monoclonal Antibody)
SDP
Supplier:  MBL International Corporation D0323
Encompass_Preferred
Clonality:Monoclonal; Application:IHC|IP|WB; Conjugate:Unlabeled; Species Reactivity:Bovine|Human|Mouse|Rat; Size:100µg; Target:Proton pump; Monoclonal antibody of 100 µg targeting Proton pump (H,K-ATPase β subunit) for IHC, IPP, WB.Proton pump (H,K-ATPase β subunit) Monoclonal Antibody.
Catalog No. 50-458-31
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.