Learn More
Selleck Chemical LLC Protoporphyrin IX 5mg 553-12-8 PPIX
SDP
Supplier:  Selleck Chemical LLC S51455MG
Encompass_Preferred
Protoporphyrin IX (PPIX) is a heterocyclic organic compound, which consists of four pyrrole rings, and is the final intermediate in the heme biosynthetic pathway. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8307
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.