Learn More
CPC Scientific FITC-LC-Gly-Arg-Gly-Asp-Ser-Pro-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific THRO004A
Encompass
This is a fluorescent (FITC)-labeled Prototype of RGD-containing peptide, Abs/Em=492/516 nm.ONE-LETTER SEQUENCE: FITC-LC-GRGDSPMOLECULAR FORMULA: C49H59N11O16SMOLECULAR WEIGHT:1090.12STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [2022956-44-9], SYNONYMS: FITC-eplison-Ahx-GRGDSP, FITC-eplison-Ahx-Gly-Arg-Gly-Asp-Ser-Pro-OHRESEARCH AREA: HematologyREFERENCES: R. Hart and D.R. Greaves, J. Immunol., 185, 3728 (2010); L. C. Zanetti-Domingues et al., PLoS One, 7, e45655 (2012)
Catalog No. 50-211-0565
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.