Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Protozoa & Algae Slide Set
SDPSCI_ED

Catalog No. S89559 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89559 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

Catalog No. S89559 Supplier United Scientific™ Supplier No. UNSLPRAL20
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prop 65 Warning: Cancer and reproductive harm

United Scientific™ Protozoa & Algae Slide Set is a collection of slides specific to the study of protozoa and algae. Set of 20.

  • Glass prepared microscope slides, supplied in a plastic case. A collection of slides specific to the study of protozoa and algae.
  • Set of 20 includes:
    • Amoeba, E
    • Mixed foraminifera
    • Mixed radiolaria
    • Euglena, E
    • Trypanosoma in blood smear
    • Plasmodium sp. (Malaria) in human blood
    • Paramecium, E
    • Paramecium showing conjugation
    • Vorticella, E
    • Chlamydomonas, E
    • Pandorina, E
    • Volvox, with daughter colonies, E
    • Nostoc, filaments with heterocysts, E
    • Cladophora, E
    • Chlorella, E
    • Spirogyra, vegetative filaments, E
    • Spirogyra, conjugation, E
    • Desmids, mixed species, E
    • Diatoms, mixed species, E
    • Paramecium fission

Specifications

Grade 5 to 12
Format Kit
Product Type Slide Set
Includes Set of 20
Languages English
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.