Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High-Tech Conversions ProWorks™ Nitrile Gloves

Catalog No. 17444087
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-444-087 Large
17-444-085 Small
17-444-086 Medium
17-444-088 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-444-087 Supplier High-Tech Conversions Supplier No. GLN135FL
Add to Cart
Add to Cart

ProWorks™ Nitrile Gloves are powder free nitrile exam gloves suitable for healthcare, laboratory, or industrial use.

  • Excellent comfort and feel
  • Textured fingers
  • Standard length with beaded cuff
  • Non-sterile

Specifications

Certifications/Compliance ASTM D 6319, FDA 21
Chemotherapeutic Rated Yes
Chlorinated No
Accelerator Free No
Applications Healthcare, Laboratory, Industrial
AQL 2.5
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 5 mil
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4 mil
Powdered Powder Free
Size Large
Thickness (English) 5 mil
Thickness (Metric) 0.12 mm
FDA 510 (k) Yes
Quantity 100/box, 10 boxes/case
Length (English) 9 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.