Description
- Aluminized PBI™/Aramid Fiber knit ARFF glove offers reflective/radiant heat protection on the top, sides, and in between the fingers
- Steamblock™ insulative pad on back of hand
- Special gripping surfaces applied to the palm
Specifications
Specifications
| Certifications/Compliance | NFPA 1976-2007 |
| Material | PBI, Aramid Fiber, Leather |
| Product Type | Proximity Firefighting Gloves |
| Size | 2X-Large |
| Glove Cuff Style | Knit |
| Liner Material | Thermal |