Specifications
| Lens Coating | Anti-fog, Anti-scratch, Anti-static |
| Lens Type | H2X |
| Certifications/Compliance | ANSI Z87.1 ,CSA Z94.3-07 |
| Certifications/Compliance (Europe) | EN-166 |
| Lens Tint | Amber |
| Product Type | Safety Spectacles |
| Frame Color | Black |
| Weight (Metric) | 26.3 g |
| Lens Material | Polycarbonate |