Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PrP Monoclonal Antibody (F89/160.1.5)
GREENER_CHOICE

Catalog No. MA1750
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1750 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1750 Supplier Invitrogen™ Supplier No. MA1750
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-750 detects prion protein (PrP) protein from a variety of species that have the conserved n-IHFG-c epitope, including agriculturally important animal species such as sheep, bovine, deer, and elk. MA1-750 has been successfully used in Western blot, immunohistochemistry and ELISA procedures. By Western blot, this antibody detects a 33-35 kDa protein from normal animals and a 27-30 kDa protein which represents PrP in brain protease treated tissue extracts from infected animals. Immunohistochemical staining of PrP(Sc) in infected sheep brain with MA1-750 results in intense staining of the spongiform lesions. In immunohistochemical procedures this antibody detects only the PrP(Sc) and not PrP(C). The MA1-750 immunogen is a synthetic peptide corresponding to residues S(146) R P L I H F G S D Y E D R(159) of bovine PrP. This peptide (Cat. # PEP-051) is available for use in neutralization and control experiments. MA1-750 is known to specifically recognize a conserved epitope of the PrP(Sc) protein comprising the amino acids n-IHFG-c.

Human prion protein (PrP), also known as PRNP, is a ubiquitously expressed GPI-anchored cell surface glycoprotein associating with lipid raft components and functioning as a signaling molecule. PrP plays a role in apoptosis in a cell context-dependent manner, is involved in proliferation of epithelial cells and in distribution of junction-associated proteins in human enterocytes. Conversion of this normal cellular prion protein (PrPc) into an abnormal conformer (PrPSc) is the crucial step associated with triggering the pathogenesis of the prion neurodegenerative disorders, such as the Creutzfeld-Jakob disease (CJD). Whereas PrPc is rich in alpha-helices, the PrPSc form has higher content of beta-sheets and is resistant to proteinase K.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PrP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone F89/160.1.5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene PRNP
Gene Accession No. F7VJQ1, P04156, P04925, P10279, P23907
Gene Alias AA960666; AI325101; Alternative prion protein; alternative prion protein; major prion protein; AltPrP; ASCR; BSE; CD230; CD230 antigen; CJD; GSS; KURU; major prion protein; major prion protein; alternative prion protein; major scrapie-associated fibril protein 1; MGC26679; p27-30; PK resistant core; prion protein; prion protein (p27-30) (Creutzfeldt-Jakob disease, Gerstmann-Strausler-Scheinker syndrome, fatal familial insomnia); prion protein precursor; prion protein precursor PrP; prion protein PrP; prion protein variant a; prion protein variant b; prion protein, PrP; prion protein, structural; prion-related protein; PRIP; Prn; Prn-i; Prnp; Prn-p; PrP; PrP>C>; PrP27-30; PrP33-35C; PrPc; PrPSc; Sinc; SIP; TSE
Gene Symbols PRNP
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues S(146) R P L I H F G S D Y E D R(159) of bovine PrP.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19122, 281427, 493887, 5621
Target Species Human, Mouse, Bovine, Sheep, Cervidae
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.