Learn More
Invitrogen™ PRP8 Recombinant Rabbit Monoclonal Antibody (JG57-38)

Description
Positive Control: A549, 293T, LOVO, SiHa, rat brain tissue, human colon tissue, human prostate cancer tissue, mouse testis tissue.
Specifications
Specifications
| Antigen | PRP8 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | JG57-38 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4 |
| Gene | Prpf8 |
| Gene Accession No. | Q6P2Q9 |
| Gene Alias | 220 kDa U5 snRNP-specific protein; apoptosis-regulated protein 1; apoptosis-regulated protein 2; AU019467; D11Bwg0410e; DBF3/PRP8; H PRPC8; HPRP8; id:ibd1257; ik:tdsubc_2a9; im:7141966; p220; precursor mRNA processing protein; pre-mRNA processing factor 8; pre-mRNA-processing-splicing factor 8; Prp8; PRP8 homolog; PRP8 pre-mRNA processing factor 8 homolog; PRPC8; prpf8; RP13; Sfprp8l; SNRNP220; spl; Splicing factor Prp8; tdsubc_2a9; U5 snRNP-specific protein; U5 snRNP-specific protein (220 kD), ortholog of S. cerevisiae Prp8p; wu:fb37c02; wu:fb73e06; xx:tdsubc_2a9; zgc:56504 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.