Learn More
Invitrogen™ PRPF39 Polyclonal Antibody

Description
The antibody detects endogenous levels of total PRPF39 protein.
Specifications
Specifications
| Antigen | PRPF39 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PRPF39 |
| Gene Accession No. | Q86UA1 |
| Gene Alias | flj11128; flj20666; flj45460; mgc149842; mgc149843; pre-mRNA processing factor 39; pre-mRNA-processing factor 39; PRP39 homolog; PRP39 pre-mRNA processing factor 39 homolog; PRP39 pre-mRNA processing factor 39 homolog (yeast); PRPF39; Srcs1; v-src suppressed transcript 1 |
| Gene Symbols | PRPF39 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.