Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRR13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565233
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565233 Supplier Invitrogen™ Supplier No. PA565233
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PYPPPAPGIPP VNPLAPGMVG PAVIVDKKMQ KK Highest antigen sequence identity to the following orthologs - mouse 73%, rat 70%.

Negatively regulates TSP1 expression at the level of transcription. This down-regulation was shown to reduce taxane-induced apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRR13
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRR13
Gene Accession No. Q9NZ81
Gene Alias BM-041; proline rich 13; proline-rich protein 13; PRR13; Taxane-resistance protein; TXR1
Gene Symbols PRR13
Host Species Rabbit
Immunogen Recombinant Human PRR13. Recombinant protein control fragment (Product #RP-97899).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54458
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.