Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRRS Virus M Protein (Strain: VR-2322) Recombinant Rabbit Monoclonal Antibody (HL2575)
GREENER_CHOICE

Catalog No. MA556847
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA556847 Supplier Invitrogen™ Supplier No. MA556847
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

PRRS Virus M Protein (Strain: VR-2322) Recombinant Monoclonal Antibody for Western Blot

Porcine reproductive and respiratory syndrome virus (PRRSV) is a highly infectious pathogen that causes severe diseases in pigs and great economic losses to the swine industry worldwide. GP5 and M are the major envelope proteins encoded by PRRSV.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRRS Virus M Protein (Strain: VR-2322)
Applications Western Blot
Classification Recombinant Monoclonal
Clone HL2575
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Porcine reproductive and respiratory syndromevirus
Host Species Rabbit
Immunogen Synthetic peptide of PRRS virus M protein. (Strain: VR-2322).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.