Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRRS Virus Nucleocapsid Protein Recombinant Rabbit Monoclonal Antibody (HL1901)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547214
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Betaarterivirus suid 1, formerly (PRRS, Porcine reproductive and respiratory syndrome virus), is a virus that causes a disease of pigs, called porcine reproductive and respiratory syndrome, also known as blue-ear pig disease. The Nucleocapsid protein is an internal structure of the virus that comprised of the association of the N protein and the RNA genome.
Specifications
| PRRS Virus Nucleocapsid Protein | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL1901 | |
| Unconjugated | |
| Porcine reproductive and respiratory syndrome virus ORF7; PRRSV ORF7 | |
| Full length PRRS virus Nucleocapsid protein. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction