Learn More
ATCC pRS314 phagemid; in E. coli
Supplier:  ATCC 77143
Encompass
One of a series of pBluescript-based centromere vectors (ATCC 77142 -77145, 77157-77158) differing in the yeast selectable marker gene.  YC-type centromere vector permitting visual detection of recombinants and production of ssDNA in E. coli. Contains promoters for in vitro RNA synthesis, priming sites useful for sequencing, and encodes the lacZ alpha (lacZ?) peptide. pRSS56, constructed by ligating a PvuI fragment (bp 498-2412) of pBluescript KS+ to a PvuI fragment (bp 2850 -730) of pBS(+), contains the KS MCS from pBluescript KS(+) and the unique NdeI and AatII sites between bla and f1 origin of pBS(+). A genomic HindII/PstI fragment (1.002 kb) containing the TRP1 gene was inserted into the NdeI site and a cassette containing CEN6 and the ARS associated with histone 4 (ARSH4) was inserted into the AatII site of pRSS56.  All ends were blunted. An EcoRI site in the TRP1-containing fragment (external to the coding sequence) was destroyed.
Catalog No. NC2327208
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.