Learn More
Postnova Analytics PS NIST TRACEABLE SIZE 200NM
SDP
Supplier:  Postnova Analytics DS3200A
Encompass
PS NIST traceable size standard 200 nm, 1.7%CV, 1% solid, 15 mL
Catalog No. NC1931027
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.