Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ pS2/pNR-2 Estrogen-Regulated Protein Ab-2 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. ms199r1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
Concentration:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS199R1 1 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS199R1 Supplier Epredia™ Supplier No. MS199R1

Provide accurate, reproducible results with the Epredia™ pS2/pNR-2 Estrogen-Regulated Protein Ab-2, Mouse Monoclonal Antibody.

pS2 is a cysteine-rich, 6.5kDa protein found in both estrogen-dependent (breast tumors) and estrogen-independent tissues (normal stomach mucosa). About 60% of breast carcinomas are positive for pS2. Staining is cytoplasmic, often with localization to the Golgi apparatus. pS2 is primarily expressed in estrogen receptor-positive breast tumors.

Host Species: Mouse

Clone: GE2; same as R47/94

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: aa 57-84

Immunogen: A synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCF-YPNTIDVPPEEECEF (aa 57-84) from C-terminus of human pS2 protein

Molecular Weight: 6.5kDa

Positive Control: Normal stomach or breast CA

Cellular Localization: Cytoplasmic Antibody to pS2 is reportedly useful in identifying a subset of estrogen-dependent breast tumors which may respond to endocrine therapy.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Affinity Purification

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen pS2/pNR-2 Estrogen-Regulated Protein Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GE2; previously R47/94
Concentration Prediluted
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen A Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCF-YPNTIDVPPEEECEF (aa 57-84) from C-terminus of human pS2 protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.