Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam PS48, Allosteric PDK1 Activator, DMSO Solution, 5 mg

Catalog No. 502704544
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4544 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4544 Supplier Abcam Supplier No. AB1466265MG
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allosteric PDK1 activator that stimulates PI3-kinase and phosphorylates AKT.

  • Molecular Weight: 286.8 Da
  • Purity: >95%
  • Mechanism: Allosteric PDK1 activator; binds to the PIF pocket in the small lobe of PDK1
  • Biological Activity: Stimulates PI3-kinase and phosphorylates AKT; shows developmental effects in vivo

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule PDK1
Substrate Properties Chemical Substrate
Buffer DMSO
Format Vial
Form Solution
Formulation In DMSO
Quantity 5 mg
For Use With (Application) PI3-kinase stimulation, AKT phosphorylation, PDK1 activation
Storage Requirements Store at -20°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.