Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSCA Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980432
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0432 Supplier Bioss Supplier No. BS0669R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSCA (Prostate Stem Cell antigen) is a cell surface antigen, which is overexpressed in approximately40% of primary prostate cancers and in as many as 100% of metastatic ones. PSCA is also overexpressed in a majority of transitional cell and pancreatic carcinomas. Antibody directed against PSCA inhibits tumorigenesis, slows tumor growth, prolongs survival and prevents metastasis in a preclinical zenograft model indicating that PSCA may have utility as a prognostic marker and/or therapeutic target in prostate cancer.

Specifications

Antigen PSCA
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PSCA
Gene Accession No. O43653
Gene Alias 2210408B04Rik; caldesmon; CDM; HCAD; H-CAD; LCAD; L-CAD; NAG22; PRO232; Prostate stem cell antigen; Psca; UNQ206; UNQ206/PRO232
Gene Symbols PSCA
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PSCA.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8000
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.