Learn More
CPC Scientific H-Lys-Asp-Thr-Lys-Pro-Thr-Pro-Thr-Lys-Gly-Lys-Ala-Ala-Ser(PO3H2)-Ala-Ser-Thr-Thr-Pro-Val-Lys-Lys-Asp-Val-Thr-Pro-Val-Lys-Ala-Asp-Thr-OH (trifluoroacetate salt) 0.25MG
SDP
Supplier:  CPC Scientific HIST009A
Encompass
Phosphorylation at histone H1 Ser42 occurs only after phosphorylation at Thr34, Thr46, or Thr53 in vivo. This peptide is histone H1 (29-59) phosphorylated at Ser42. Histone H1 phosphorylation is associated with chromosome condensation and transcriptional regulation.ONE-LETTER SEQUENCE: KDTKPTPTKGKAA-pS-ASTTPVKKDVTPVKADTMOLECULAR FORMULA: MOLECULAR WEIGHT:3249.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Garcia, B. et al. Mol Cell Proteomics 5, 1593 (2006) ; Dou, Y. and MA. Gorovsky Mol Cell 6, 225 (2000) ; Mizzen, C. et al. J Biol Chem 274, 14533 (1999); Swank, R. et al. Biochem 36, 13761 (1997)
Catalog No. 50-210-9438
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.