Learn More
Montana Molecular Pseudo SARS-CoV-2 Delta Variant Green Reporter - 5mL
SDP
Supplier:  Montana Molecular C1123G005
Encompass_Preferred
This pseudovirus is a BSL-1 baculovirus vector pseudotyped with Spike proteins, in this case the B.1.617.2 Delta Variant. Biologists and drug discovery teams can use this reagent to work on COVID-19 projects outside of BSL-3 facilities with standard safety procedures. Once inside the cell, the pseudovirus does not replicate, but delivers a genetically-encoded, fluorescent reporter that expresses bright green fluorescence in the host cell nucleus. If needed, cellular entry can be facilitated with ACE2 and other proteins packaged in BacMam and available from Montana Molecular. Larger sizes available upon request. This kit includes the pseudovirus and 500mM sodium butyrate in H2O.
Catalog No. NC2600306
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.