Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pseudomonas aeruginosa Monoclonal Antibody (B11)
GREENER_CHOICE

Catalog No. MA183430 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA183430 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA183430 Supplier Invitrogen™ Supplier No. MA183430
  • $663.00
    $506.65 / Each of 1

    Save $156.35

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Mouse anti Pseudomonas aeruginosaantibody, clone B11 recognizes Pseudomonas aeruginosa a gram-negative, opportunistic pathogen, commonly found in soil and water.

Pseudomonas Aeruginosa is a gram-negative, opportunistic pathogen commonly found in soil and water. In humans, P. aeruginosa can infect the urinary tract, respiratory and gastrointestinal system, soft tissues, bones and joints leading to severe systemic infections of immunosuppressed hospitalized patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pseudomonas aeruginosa
Applications ELISA
Classification Monoclonal
Clone B11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide; pH 7.2
Gene Alias p aeruginosa; P. aeruginosa
Host Species Mouse
Immunogen Purified outer membrane protein of Pseudomonas aeruginosa.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.