Learn More
Description
- Simply rehydrate LYFO DISK qualitative microorganism pellets with a diluent, such as phosphate buffer or saline to inoculate culture media.
- Packaged in a glass vial containing 6 pellets of a single strain
Specifications
Specifications
| ATCC® | ATCC 17386 |
| Biosafety Level | 1 |
| Content And Storage | 2°C to 8°C |
| Format | Lyfo Disk |
| Manufacturing Origin | United States |
| Product Type | Quality Control |
| For Use With (Application) | Microbial Identification |
| Packaging Type | Plastic Canister |
| Quantity | 6 Pellets |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
