Learn More
Creative Diagnostics Pseudotyped Luciferase rSARS-CoV-2 Spike
Supplier:  Creative Diagnostics COVPS01
Encompass
Pseudotyped Luciferase rSARS-CoV-2 Spike is a replication-restricted, recombinant pseudotyped lentiviral particles containing SARS-CoV-2 spike protein. Because the infectivity of Pseudotyped Luciferase rSARS-CoV-2 is restricted to a single round of replication, the pseudotypes can be handled using BSL-2 containment practices.
Catalog No. NC1812191
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.