Learn More
AdipoGen Pseudouridimycin
SDP
Supplier:  AdipoGen AGCN20316M001
Encompass_Preferred
Chemical. CAS 1566586-52-4. Formula C17H26N8O9. MW 486.4. Isolated from Streptomyces sp. Peptidyl nucleoside antibiotic. Selective nucleoside-analog inhibitor of bacterial RNA polymerase RNAP. Inhibits RNAP through a distinctive binding site on RNAP the NTP addition site and mechanism competition with UTP for occupancy of the NTP addition site than Rifampin. Targets bacterial RNAP in Gram-positive and Gram-negative organisms, including antibiotic-resistant isolates and shows extremely low frequency of resistance selection. Potently inhibits bacterial RNAP in vitro, inhibits bacterial growth in culture, and has shown in vivo efficacy in animal models of infection.
Catalog No. 50-256-2607
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.