Learn More
Cytiva (Formerly Pall Lab) Acrodisc PSF Syringe Filter with Supor Membrane - GxF/0.45 m, 25 mm (1000/pkg)
Supplier:  Cytiva AP4426
Encompass_Preferred
Acrodisc PSF Syringe Filter with Supor Membrane - GxF/0.45 m, 25 mmSyringe filters with Supor (polyethersulfone) membrane are ideal for the filtration of aqueous and proteinaceous solutions in HPLC (0.45 m) and UHPLC (0.2 m) sample preparation. Sample filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending chromatography column life and reducing system downtime.Acrodisc PSF Syringe Filters have a large effective filtration area (3.9 cm2) for increased sample throughput, ideal for sample volumes of 150 mL, and features a built-in GxF prefilter for use with difficult-to-filter solutions.The syringe filters are designed to be fully compatible with automated robotic handling systems. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3405
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.