Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSG11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559926
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559926 Supplier Invitrogen™ Supplier No. PA559926
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STSLTIRVIA PPGLGTFAFN NPT Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 43%.

PSG11, also named as PSG13, PSG14, PSBG-11 and PSBG-13, belongs to the immunoglobulin superfamily and CEA family. PSBG are produced in high quantity during pregnancy. PSG11 and other PSGs induce dose-dependent secretion of anti-inflammatory cytokines by human monocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSG11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PSG11
Gene Accession No. Q9UQ72
Gene Alias pregnancy specific beta-1-glycoprotein 11; pregnancy-specific beta-1-glycoprotein 11; Pregnancy-specific beta-1-glycoprotein 13; PS-beta-G-11; PS-beta-G-13; PSBG-11; PSBG-13; PSG11; PSG13; PSG14
Gene Symbols PSG11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PSG11. Recombinant protein control fragment (Product #RP-99055).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5680
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.