Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598411
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598411 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598411 Supplier Invitrogen™ Supplier No. PA598411
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSG3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The pregnancy-specific glycoproteins (PSGs) are a group of closely related secreted glycoproteins that are highly expressed in fetal placental syncytiotrophoblast cells. Members of the PSG protein family share a characteristic N-terminal domain that is homologous to the immunoglobulin variable region. PSGs become detectable in serum during the first two to three weeks of pregnancy and increase as the pregnancy progresses, eventually representing the most abundant fetal protein in the maternal blood at term. PSGs function to stimulate secretion of TH2-type cytokines from monocytes, and they may also modulate the maternal immune system during pregnancy, thereby protecting the semi-allotypic fetus from rejection. Eleven human PSG proteins (PSG1-PSG11) have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSG3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PSG3
Gene Accession No. Q16557
Gene Alias Carcinoembryonic antigen SG5; pregnancy specific beta-1-glycoprotein 3; pregnancy-specific beta-1-glycoprotein 3; pregnancy-specific glycoprotein 3; PS-beta-G-3; PSBG-3; PSG3; sg5
Gene Symbols PSG3
Host Species Rabbit
Immunogen Recombinant Human Pregnancy-specific beta-1-glycoprotein 3 protein (35-310AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5671
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.