Learn More
Description
This antibody recognizes extracellular domain (preferentially in native form) of glutamate carboxypeptidase II (NAALADase, FOLH1, PSMA), an approximately 95-110 kDa transmembrane glycoprotein expressed mainly in tumor neovasculatures, nervous system and jejunum, which is an important prostate tumor marker.
Specifications
Specifications
| Antigen | PSMA |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | GCP-05 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 |
| Gene | FOLH1 |
| Gene Accession No. | Q04609 |
| Gene Alias | Cell growth-inhibiting gene 27 protein; FGCP; folate hydrolase (prostate-specific membrane antigen) 1; folate hydrolase 1; FOLH; Folh1; Folylpoly-gamma-glutamate carboxypeptidase; GCP2; GCPII; GIG27; glutamate carboxylase II; glutamate carboxypeptidase 2; Glutamate carboxypeptidase II; membrane glutamate carboxypeptidase; mGCP; mopsm; Naalad; NAALAD1; NAALAdase; NAALADase I; N-acetylated alpha-linked acidic dipeptidase; N-acetylated alpha-linked acidic dipeptidase 1; N-acetylated-alpha-linked acidic dipeptidase I; prostate specific membrane antigen; prostate specific membrane antigen variant F; prostate-specific membrane antigen; Prostate-specific membrane antigen homolog; PSM; PSMA; pteroylpoly-gamma-glutamate carboxypeptidase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
