Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMAL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524990
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524990 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524990 Supplier Invitrogen™ Supplier No. PA524990
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSMAL Polyclonal Antibody for Western Blot, IHC (P), Flow

PSMAL has both folate hydrolase and N-acetylated-alpha-linked-acidic dipeptidase (NAALADase) activity. PSMAL exhibits a dipeptidyl-peptidase IV type activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMAL
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FOLH1B
Gene Accession No. Q9HBA9
Gene Alias cell growth-inhibiting gene 26 protein; folate hydrolase 1B; folate hydrolase 2; folate hydrolase pseudogene; FOLH1B; FOLH2; FOLHP; GCP3; GCPIII; GIG26; glutamate carboxypeptidase III; NAALADase; N-acetylated-alpha-linked-acidic dipeptidase; Prostate-specific membrane antigen-like protein; PSM; PSMAL; PSMA-LIKE; Putative folate hydrolase 1B; putative N-acetylated-alpha-linked acidic dipeptidase
Gene Symbols FOLH1B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 155-183 amino acids from the central region of human PSMAL.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219595
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.