Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142607
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142607 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142607 Supplier Invitrogen™ Supplier No. PA5142607
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMB4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PSMB4
Gene Accession No. P28070, P99026
Gene Alias 26 kDa prosomal protein; HN3; HsBPROS26; HsN3; Lmp3; Low molecular mass protein 3; macropain beta chain; Multicatalytic endopeptidase complex beta chain; PROS26; PROS-26; Pros-27; proteasome (prosome, macropain) subunit, beta type 4; proteasome (prosome, macropain) subunit, beta type, 4; proteasome beta 4 subunit; proteasome beta chain; Proteasome chain 3; proteasome subunit beta 4; proteasome subunit beta type-4; proteasome subunit HsN3; proteasome subunit, beta type, 4; Psmb4; RN3; testis tissue sperm-binding protein Li 79P
Gene Symbols PSMB4
Host Species Goat
Immunogen Peptide with sequence C-QIATVTEKGVE.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19172, 5692
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.