Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMB7 Monoclonal Antibody (OTI4G9)
GREENER_CHOICE

Catalog No. PIMA525900
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA525900 100 μL
1 options

Catalog No. PIMA525900

Supplier: Invitrogen™ MA525900

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMB7 Monoclonal Antibody for Western Blot, ICC/IF

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit in the proteasome. Expression of this catalytic subunit is downregulated by gamma interferon and proteolytic processing is required to generate a mature subunit. This subunit is not present in the immunoproteasome and is replaced by catalytic subunit 2i (proteasome beta 10 subunit).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMB7
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI4G9
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Psmb7
Gene Accession No. P70195, Q99436, Q9JHW0
Gene Alias AU020723; beta-type proteasome 7 subunit; EGK_07890; hypothetical protein LOC511207; macropain chain Z; MC14; Mmc14; Multicatalytic endopeptidase complex chain Z; proteasome (prosome, macropain) subunit, beta type 7; proteasome (prosome, macropain) subunit, beta type, 7; proteasome 20S subunit beta 7; proteasome beta 7 subunit; proteasome catalytic subunit 2; proteasome subunit alpha; proteasome subunit beta 7; proteasome subunit beta type-7; proteasome subunit beta type-7 proprotein; proteasome subunit Z; protein serine kinase c17; Psmb7; RP11-101K10.7; Z
Gene Symbols Psmb7
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 58-277 of PSMB7 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100686423, 19177, 5695, 85492
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.