Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMC2 Monoclonal Antibody (1851CT193.4.40)
GREENER_CHOICE

Catalog No. PIMA537762
Change view
Click to view available options
Quantity:
200 μL
Catalog No. Quantity
PIMA537762 200 μL
1 options

Catalog No. PIMA537762

Supplier: Invitrogen™ MA537762

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMC2 Monoclonal Antibody for Western Blot

Proteolytic degradation is critical to the maintenance of appropriate levels of short-lived and regulatory proteins as important and diverse as those involved in cellular metabolism, heat shock and stress response, antigen presentation, modulation of cell surface receptors and ion channels, cell cycle regulation, transcription, and signalling factors. The ubiquitin-proteasome pathway deconstructs most proteins in the eukaryotic cell cytosol and nucleus. Others are degraded via the vacuolar pathway which includes endosomes, lysosomes, and the endoplasmic reticulum. The 26S proteasome is an ATP-dependent, multisubunit (~31), barrel-shaped molecular machine with an apparent molecular weight of ~2.5 MDa. It consists of a 20S proteolytic core complex which is crowned at one or both ends by 19S regulatory subunit complexes. The 19S regulatory subunits recognize ubiquitinated proteins and play an essential role in unfolding and translocating targets into the lumen of the 20S subunit. An enzymatic cascade is responsible for the attachment of multiple ubiquitin molecules to lysine residues of proteins targeted for degradation. Several genetic diseases are associated with defects in the ubiquitin-proteasome pathway. Some examples of affected proteins include those linked to cystic fibrosis, Angelman's syndrome, and Liddle syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMC2
Applications Western Blot
Classification Monoclonal
Clone 1851CT193.4.40
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PSMC2
Gene Accession No. P35998
Gene Alias 26S protease regulatory subunit 7; 26S proteasome AAA-ATPase subunit RPT1; 26S proteasome regulatory subunit 7; mammalian suppressor of sgv-1 of yeast; MGC3004; MSS1; Nbla10058; protease 26S subunit 7; proteasome (prosome, macropain) 26S subunit, ATPase 2; proteasome (prosome, macropain) 26S subunit, ATPase, 2; proteasome 26S subunit ATPase 2; proteasome 26S subunit, ATPase 2; proteasome 26S subunit, ATPase, 2; Protein MSS1; PSMC2; putative protein product of Nbla10058; S7; testis secretory sperm-binding protein Li 197a
Gene Symbols PSMC2
Host Species Mouse
Immunogen recombinant protein from human PSMC2.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5701
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.