Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMC6 Recombinant Rabbit Monoclonal Antibody (PSH0-12)
GREENER_CHOICE

Catalog No. FEMA550776
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550776 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550776 Supplier Invitrogen™ Supplier No. MA550776
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: Hela cell lysate, 293T cell lysate, SH-SY5Y cell lysate, HepG2 cell lysate, PC-12 cell lysate, NIH/3T3 cell lysate, mouse kidney tissue lysate, rat kidney tissue lysate, rat brain tissue lysate, human testis tissue, SH-SY5Y, NIH/3T3, PC-12. Predicted band size: 44 kDa Subcellular Location: Cytoplasm, Nucleus.

PSMC6 (proteasome (prosome, macropain) 26S subunit, ATPase 6), also known as P44, p42, SUG2, S10B or CADP44, is a regulatory component of the 26S proteasome. More specifically, PSMC6 is one of the six ATPase subunits of the 19S regulator base. It contains a leucine zipper motif and an AAA (ATPase associated with diverse cellular activities) domain and belongs to the AAA ATPase family of chaperone-like ATPases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMC6
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone PSH0-12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PSMC6
Gene Accession No. P62333, P62334
Gene Alias 2300001E01Rik; 26S protease regulatory subunit 10B; 26S protease regulatory subunit S10B; 26S proteasome AAA-ATPase subunit RPT4; 26S proteasome regulatory subunit 10B; AI451058; CADP44; conserved ATPase domain protein 44; epididymis secretory protein Li 73; HEL-S-73; p42; P44; proteasome (prosome, macropain) 26S subunit, ATPase, 6; proteasome (prosome, macropain) 26S subunit, ATPase, 6-like; proteasome 26S subunit ATPase 6; proteasome 26S subunit, ATPase 6; proteasome subunit p42; PSMC6; SUG2
Gene Symbols PSMC6
Host Species Rabbit
Immunogen Recombinant protein within human PSMC6 aa 1-200/389.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289990, 5706, 67089
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.