Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1974 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1974 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1974 Supplier Invitrogen™ Supplier No. PA1974
  • $596.37
    $461.65 / Each of 1

    Save $134.72

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-974 detects proteasome 19S subunit S3 from human and mouse cells. PA1-974 has been successfully used in ICC/IF, FACS, and Western blot procedures. By Western blot, this antibody detects an ~60 kDa protein representing proteasome 19S subunit S3 from HeLa cell extract. For Western blot procedures it is necessary to transfer the protein to membrane at pH 11. PA1-974 immunizing peptide corresponds to amino acid residues 513-534 from human proteasome 19S subunit S3 protein.

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes one of the non-ATPase subunits of the 19S regulator lid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMD3
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene PSMD3
Gene Accession No. O43242, P14685
Gene Alias 26S proteasome non-ATPase regulatory subunit 3; 26S proteasome regulatory subunit RPN3; 26S proteasome regulatory subunit S3; AI255837; AntP91a; D11Bwg1349e; EGK_08646; P58; P91a; proteasome (prosome, macropain) 26S subunit, non-ATPase, 3; proteasome 26S non-ATPase subunit 3; proteasome 26S subunit, non-ATPase 3; proteasome subunit p58; Psd3; Psmd3; RPN3; S3; tissue specific transplantation antigen 2; tissue specific transplantation antigen P91A; transplantation antigen P91A; TSTA2; Tstap91a; Tum-P91A antigen
Gene Symbols PSMD3
Host Species Rabbit
Immunogen Synthetic Peptide: E(513) R E Q Q D L E F A K E M A E D D D D S F P(534).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22123, 5709
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.