Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSME3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517333
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517333 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517333 Supplier Invitrogen™ Supplier No. PA517333
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

SFRS10 contains 1 RRM (RNA recognition motif) domain and belongs to the splicing factor SR family. It is a sequence-specific RNA-binding protein which participates in the control of pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSME3
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 6 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene PSME3
Gene Accession No. P61289, P61290
Gene Alias 11S regulator complex gamma subunit; 11S regulator complex subunit gamma; AA410043; Ab2-371; Activator of multicatalytic protease subunit 3; AU020960; epididymis secretory protein Li 283; HEL-S-283; Ki; Ki antigen; Ki nuclear autoantigen; PA28 gamma variant 5; pa28g; PA28gamma; PA28-gamma; prosome macropain; proteaseome (prosome, macropain) 28 subunit, 3; proteaseome (prosome, macropain) activator subunit 3 (PA28 gamma, Ki); proteasome (prosome, macropain) activator subunit 3; proteasome (prosome, macropain) activator subunit 3 (PA28 gamma; Ki); proteasome activator 28 subunit gamma; proteasome activator 28-gamma; proteasome activator complex subunit 3; proteasome activator subunit 3; Psme3; REG gamma-3; REGgamma; REG-gamma
Gene Symbols PSME3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding Asp16 of human PA2 gamma.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10197, 19192, 287716
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.