Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMF1 Monoclonal Antibody (OTI2D7)
GREENER_CHOICE

Catalog No. PIMA526099
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA526099 100 μL
1 options

Catalog No. PIMA526099

Supplier: Invitrogen™ MA526099

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMF1 Monoclonal Antibody for Western Blot, ICC/IF

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a protein that inhibits the activation of the proteasome by the 11S and 19S regulators. Alternative transcript variants have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMF1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PSMF1
Gene Accession No. Q92530
Gene Alias AW048666; BC012260; hPI31; PI31; proteasome (prosome, macropain) inhibitor subunit 1; proteasome (prosome, macropain) inhibitor subunit 1 (PI31); proteasome inhibitor PI31 subunit; proteasome inhibitor subunit 1; PSMB2; Psmf1; RP23 402M7.5; RP4 545L17.1; RP4-545L17.1
Gene Symbols PSMF1
Host Species Mouse
Immunogen Full length human recombinant protein of PSMF1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9491
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.